Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD36/SR-B3 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB25191V100UG
This item is not returnable.
View return policy
Description
CD36/SR-B3 Monoclonal specifically detects CD36/SR-B3 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD36/SR-B3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q08857 | |
CD36 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD36/SR-B3 Gly30-Lys439 Accession # Q08857 | |
RUO | |
948 | |
Mouse | |
IgG1 |
Flow Cytometry | |
324205 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, Collagen R, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, SRB3, SR-B3, Thrombospondin R, Thrombospondin receptor | |
Rat | |
100 μg | |
Primary | |
Detects mouse CD36/SR-B3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CD36 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction