Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD36/SR-B3 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA25191R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA25191R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA25191R100 Supplier R&D Systems Supplier No. FAB25191R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD36/SR-B3 Monoclonal specifically detects CD36/SR-B3 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD36/SR-B3
Applications Flow Cytometry
Classification Monoclonal
Clone 324205
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q08857
Gene Alias BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, Collagen R, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, SRB3, SR-B3, Thrombospondin R, Thrombospondin receptor
Gene Symbols CD36
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD36/SR-B3 Gly30-Lys439 Accession # Q08857
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 948
Test Specificity Detects mouse CD36/SR-B3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CD36 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.