Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD38 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4947
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4947 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4947 Supplier R&D Systems Supplier No. BAF4947
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CD38 Polyclonal specifically detects CD38 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CD38
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. EDL37596
Gene Alias 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl Cyclase, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose Hydrolase, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10
Gene Symbols CD38
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD38 Leu45-Thr304 Accession # EDL37596
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 952
Test Specificity Detects mouse CD38 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human CD38 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.