Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD39/ENTPD1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4398T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4398T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4398T100 Supplier R&D Systems Supplier No. FAB4398T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD39/ENTPD1
Applications Flow Cytometry
Classification Monoclonal
Clone 495826
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAH11278
Gene Alias ADPase, ATPDase, ATP-diphosphatase, CD39, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, ENTPD1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPD1, NTPDase 1, NTPDase-1
Gene Symbols ENTPD1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD39/ENTPD1 Thr38-Ile478 Accession # AAH11278
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 953
Test Specificity Detects mouse CD39/ENTPD1 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human CD39 is observed and no cross-reactivity with recombinant mouse CD39L3 or recombinant human CD39L4 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.