Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD39/ENTPD1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB4398SP
This item is not returnable.
View return policy
Description
CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready.Specifications
CD39/ENTPD1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready | |
AAH11278 | |
ENTPD1 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD39/ENTPD1 Thr38-Ile478 Accession # AAH11278 | |
25 μg | |
Primary | |
Detects mouse CD39/ENTPD1 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human CD39 is observed and no cross-reactivity with recombinant mouse CD39L3 or recombinant human CD39L4 is observed. | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
495826 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ADPase, ATPDase, ATP-diphosphatase, CD39, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, ENTPD1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPD1, NTPDase 1, NTPDase-1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
953 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction