Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD4 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

Specifications

Antigen CD4
Clone GK1.5
Dilution Flow Cytometry 5 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB554V025
GREENER_CHOICE
View Documents
R&D Systems
FAB554V025
25 Tests N/A N/A N/A
FAB554V100
GREENER_CHOICE
View Documents
R&D Systems
FAB554V100
100 Tests N/A N/A N/A
Description

Description

CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD4
Flow Cytometry 5 uL/10^6 cells
Monoclonal
Supernatant
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
920
Mouse CTL clone V4
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
GK1.5
Flow Cytometry
Alexa Fluor 405
Rat
Mouse
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
CD4
IgG2b
Protein A or G purified from hybridoma culture supernatant
Reacts with mouse CD4, an antigen co-receptor on the Tácell surface which interacts with MHCáII molecules on antigen presenting cells.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.