Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD4 Fluorescein-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems FAB554F100
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD4 | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD4 | |
Mouse CTL clone V4 | |
100 Tests | |
Primary | |
Reacts with mouse CD4, an antigen co-receptor on the Tácell surface which interacts with MHCáII molecules on antigen presenting cells. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Flow Cytometry | |
GK1.5 | |
Flow Cytometry 10 uL/10^6 cells | |
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
920 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction