Learn More
Mouse CD40 Ligand/TNFSF5 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CD40 Ligand/TNFSF5 |
| Applications | Western Blot, Neutralization, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Neutralization 0.08-0.4 ug/mL, Immunocytochemistry 5-15 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P27548 |
| Gene Alias | CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40L, CD40-L, CD40LG, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5, TNFSF5IMD3, TRAP, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 |
| Gene Symbols | CD40LG |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.