Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD45 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB114R025UG
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD45 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 0.25-1 μg/106 cells | |
B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse CD45. Recognizes all CD45 isoforms as well as the known mouse CD45 alloantigens (CD45-1/Ly 5a and CD45-2/Ly 5b) (15). | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Flow Cytometry | |
30-F11 | |
Alexa Fluor 647 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
PTPRC | |
Mouse thymus and spleen cells | |
25 μg | |
5788.0 | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction