Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD47 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB18661U10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB18661U10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB18661U10 Supplier R&D Systems Supplier No. FAB18661U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD47 Monoclonal antibody specifically detects CD47 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen CD47
Applications Flow Cytometry
Classification Monoclonal
Clone 974222
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. NP_034711
Gene Alias antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAP, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived mouse CD47, Gln19-Pro158, Accession # NP_034711
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 961
Reconstitution Reconstitution default not known
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.