Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD47 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $444.00
Specifications
Antigen | CD47 |
---|---|
Clone | 974222 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 0.08-0.8 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Blocking Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB18661SP
![]() |
R&D Systems
MAB18661SP |
25 μg |
Each for $200.50
|
|
|||||
MAB18661100
![]() |
R&D Systems
MAB18661100 |
100 μg |
Each for $444.00
|
|
|||||
Description
CD47 Monoclonal specifically detects CD47 in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
CD47 | |
Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 0.08-0.8 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAP, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
Mouse myeloma cell line NS0-derived mouse CD47, Gln19-Pro158, Accession # NP_034711 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
974222 | |
Flow Cytometry, Blocking Assay, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
NP_034711 | |
961 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title