Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD48/SLAMF2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3327
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3327 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3327 Supplier R&D Systems Supplier No. AF3327
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CD48/SLAMF2 Polyclonal specifically detects CD48/SLAMF2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CD48/SLAMF2
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. BAE96326
Gene Alias BCM1, BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Gene Symbols CD48
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD48/SLAMF2 Phe23-Arg216 Accession # BAE96326
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 962
Test Specificity Detects mouse CD48/SLAMF2 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human CD48, recombinant mouse (rm) OX40, rm2B4, and rmNTB-A, and rmCD2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.