Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Mouse CD48/SLAMF2 ELISA Kit (Colorimetric)
SDP

Catalog No. NB231923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB231923 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB231923 Supplier Novus Biologicals Supplier No. NBP280373

Specifications

Assay Range 62.5-4000 pg/mL
Assay Sensitivity 16.01 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Lysates
Specificity Recognizes both recombinant and natural Mouse CD48/SLAMF2.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 962
Gene Symbol CD48
Quantity 1 Kit
Research Discipline Cell Biology, Cellular Markers, Immunology, Stem Cell Markers
Storage Requirements Storage is content dependent.
Synonym BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.