Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD55/DAF Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5376G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5376G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5376G100 Supplier R&D Systems Supplier No. FAB5376G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD55/DAF Monoclonal specifically detects CD55/DAF in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD55/DAF
Applications Flow Cytometry
Classification Monoclonal
Clone 583905
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q61475
Gene Alias CD55, CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CR, CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, DAF, decay accelerating factor for complement, TC
Gene Symbols CD55
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD55/DAF Met1-Pro359 Accession # Q61475
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1604
Test Specificity Detects mouse CD55/DAF in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CD55 or recombinant mouse CD97 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.