Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD6 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB727S100UG
This item is not returnable.
View return policy
Description
CD6 Monoclonal specifically detects CD6 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD6 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q61003 | |
CD6 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD6 Leu18-Gly396 Accession # Q61003 | |
RUO | |
923 | |
Mouse | |
IgG2a |
Flow Cytometry | |
96123 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD6, CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
Rat | |
100 μg | |
Primary | |
Detects mouse CD6 in direct ELISAs and Western blots. Does not cross-react with human CD6. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction