Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD6 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $410.00
Specifications
Antigen | CD6 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Adhesion Blockade |
Applications | Western Blot, Adhesion Blockade |
Classification | Polyclonal |
Description
CD6 Polyclonal specifically detects CD6 in Mouse samples. It is validated for Western Blot, Adhesion Blockade.Specifications
CD6 | |
Western Blot 0.1 ug/mL, Adhesion Blockade | |
Polyclonal | |
Goat | |
Mouse | |
CD6, CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
CD6 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Adhesion Blockade | |
Unconjugated | |
RUO | |
Q91WN5 | |
923 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD6 Leu18-Gly396 Accession # Q91WN5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse CD6 in direct ELISAs and Western blots. In Western blots, approximately 1% cross-reactivity with recombinant human CD6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title