Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM727
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM727 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM727 Supplier R&D Systems Supplier No. BAM727
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD6 Monoclonal specifically detects CD6 in Mouse samples. It is validated for ELISA Detection (Matched Antibody Pair).

Specifications

Antigen CD6
Applications ELISA
Classification Monoclonal
Clone 96123
Concentration LYOPH
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. Q61003
Gene Alias CD6, CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6
Gene Symbols CD6
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD6 Leu18-Gly396 Accession # Q61003
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 923
Test Specificity Detects mouse CD6 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human CD6 and recombinant mouse ALCAM is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.