Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD68/SR-D1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC10114R100UG
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CD68/SR-D1 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1, SRD1, SR-D1 | |
Chinese Hamster Ovary cell line, CHO-derived mouse CD68, Asp21-Pro282, Accession No. P31996 | |
100 μg | |
968.0 | |
Mouse | |
Supernatant |
Flow Cytometry | |
2449C | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P31996 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects mouse CD68 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction