Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD68/SR-D1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $514.50
Specifications
| Antigen | CD68/SR-D1 |
|---|---|
| Clone | 2449D |
| Dilution | Immunohistochemistry 3-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
| Applications | Immunohistochemistry, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB101141SP
![]() |
R&D Systems
MAB101141SP |
25 μg |
Each for $214.00
|
|
|||||
MB101141100
![]() |
R&D Systems
MAB101141100 |
100 μg |
Each for $514.50
|
|
|||||
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Mouse samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| CD68/SR-D1 | |
| Immunohistochemistry 3-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| P31996 | |
| 968 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 2449D | |
| Immunohistochemistry, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1, SRD1, SR-D1 | |
| Chinese Hamster Ovary cell line, CHO-derived mouse CD68, Asp21-Pro282, Accession # P31996 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CD68 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title