Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD69 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2386V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2386V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2386V100 Supplier R&D Systems Supplier No. FAB2386V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD69 Monoclonal specifically detects CD69 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD69
Applications Flow Cytometry
Classification Monoclonal
Clone 310106
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P37217
Gene Alias Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, EA-1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leu23, Leukocyte surface antigen Leu-23, MLR-3, p60
Gene Symbols CD69
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD69 Gly64-Arg199 Accession # P37217
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 969
Test Specificity Detects mouse CD69 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD69 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.