Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD69 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | CD69 |
---|---|
Clone | 310106 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells |
Applications | Western Blot, Flow Cytometry |
Description
CD69 Monoclonal specifically detects CD69 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD69 | |
LYOPH | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rat | |
Mouse | |
P37217 | |
969 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD69 Gly64-Arg199 Accession # P37217 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CD69 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD69 is observed. |
310106 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, EA-1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leu23, Leukocyte surface antigen Leu-23, MLR-3, p60 | |
CD69 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title