Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD79B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6814SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6814SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6814SP Supplier R&D Systems Supplier No. AF6814SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CD79B Polyclonal specifically detects CD79B in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen CD79B
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P15530
Gene Alias B29, B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79B, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGB, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Gene Symbols CD79B
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD79B Met29-Asp158 Accession # P15530
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 974
Test Specificity Detects mouse CD79B in direct ELISAs and Western blots. In direct ELISAs, approximately 55% of cross-reactivity with human CD79B is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.