Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD8 alpha Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
$176.63 - $354.76
Specifications
| Antigen | CD8 |
|---|---|
| Clone | 53-6.7 |
| Dilution | Flow Cytometry 5 uL/10^6 cells, Immunocytochemistry 5 ug/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB116G025
![]() |
R&D Systems
FAB116G025 |
25 Tests |
Each for $176.63
|
|
|||||
FAB116G100
![]() |
R&D Systems
FAB116G100 |
100 Tests |
Each for $354.76
|
|
|||||
Description
CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
| CD8 | |
| Flow Cytometry 5 uL/10^6 cells, Immunocytochemistry 5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 925 | |
| Mouse thymus or spleen | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 53-6.7 | |
| Flow Cytometry, Immunocytochemistry | |
| Alexa Fluor 488 | |
| Rat | |
| Mouse | |
| CD8, CD8A, CD8a molecule, LEU2, p32 | |
| CD8A | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects both the alpha and alpha ' chains of mouse CD8 (1). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title