Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD8 alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 7 publications
$227.50 - $2110.00
Specifications
Antigen | CD8 |
---|---|
Clone | 53-6.7 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation, Cell depletion, Inhibition of T Cell Function, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunoprecipitation, Depletion Assay, Inhibition Assays, Immunocytochemistry |
Classification | Monoclonal |
Description
CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Cell depletion, Inhibition of T Cell Function, Immunocytochemistry, CyTOF-ready.Specifications
CD8 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation, Cell depletion, Inhibition of T Cell Function, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
925 | |
Mouse thymus or spleen | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects both the alpha and alpha ' chains of mouse CD8 (1). |
53-6.7 | |
Flow Cytometry, Immunoprecipitation, Depletion Assay, Inhibition Assays, Immunocytochemistry | |
Unconjugated | |
Rat | |
Mouse | |
CD8, CD8A, CD8a molecule, LEU2, p32 | |
CD8A | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title