Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD9 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5218S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5218S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5218S100 Supplier R&D Systems Supplier No. FAB5218S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD9 Monoclonal specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD9
Applications Flow Cytometry
Classification Monoclonal
Clone 479608
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P40240
Gene Alias BA2, BA-2/p24 antigen, BTCC-1, CD9, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Gene Symbols CD9
Host Species Rat
Immunogen NS0 mouse myeloma cell line transfected with mouse CD9 Pro2-Val226 Accession # P40240
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 928
Test Specificity Stains mouse CD9-transfected cells but not irrelevant transfecants.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.