Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD90/Thy1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7335
This item is not returnable.
View return policy
Description
CD90/Thy1 Polyclonal specifically detects CD90/Thy1 in Mouse samples. It is validated for Western Blot.Specifications
CD90/Thy1 | |
Polyclonal | |
Unconjugated | |
P01831 | |
THY1 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD90/Thy1 Gln20-Lys130 Accession # P01831 | |
100 μg | |
Primary | |
Detects mouse CD90/Thy1 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human CD90/Thy1 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL | |
CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
Sheep | |
Affinity Purified | |
RUO | |
7070 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction