Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CDCP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4515SP
This item is not returnable.
View return policy
Description
CDCP1 Polyclonal specifically detects CDCP1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CDCP1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
Sheep | |
Affinity Purified | |
RUO | |
64866 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q5U462 | |
CDCP1 | |
Mouse myeloma cell line NS0-derived recombinant mouse CDCP1 Arg25-Ala667 Accession # Q5U462 | |
25 μg | |
Primary | |
Detects mouse CDCP1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human CDCP1 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction