Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CDNF Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5187
This item is not returnable.
View return policy
Description
CDNF Polyclonal specifically detects CDNF in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CDNF | |
Polyclonal | |
Unconjugated | |
Q8CC36 | |
CDNF | |
Mouse myeloma cell line NS0-derived recombinant mouse CDNF Leu27-Leu187 Accession # Q8CC36 | |
100 μg | |
Primary | |
Detects mouse CDNF in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human CDNF is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
arginine-rich, mutated in early stage tumors-like 1, ARMETL1, ARMET-like protein 1, cerebral dopamine neurotrophic factor, Conserved dopamine neurotrophic factor | |
Goat | |
Affinity Purified | |
RUO | |
441549 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction