Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CEACAM-1/CD66a APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6480A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6480A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6480A Supplier R&D Systems Supplier No. FAB6480A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CEACAM1/CD66a
Applications Flow Cytometry
Classification Monoclonal
Clone 723629
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P31809
Gene Alias antigen CD66, BGP1, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, Biliary Glycoprotein 1, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen, Cea-1, CEACAM1, Hv-1, Hv-2, MHVR
Gene Symbols Ceacam1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-1/CD66a Glu35-Gly428 Accession # P31809
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 634
Test Specificity Detects mouse CEACAM-1/CD66a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) CECAM-1, -3, -5, or -6 is observed. In Western blots, no cross-reactivity with rhCEACAM-1, -3, -4, -5, -6, or -7 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.