Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CEACAM-2 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. F9214R100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F9214R100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. F9214R100UG Supplier R&D Systems Supplier No. FAB9214R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CEACAM2 Monoclonal specifically detects CEACAM2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CEACAM2
Applications Flow Cytometry
Classification Monoclonal
Clone 935610
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.1 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q925P2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2 Gln35-Asp422 Accession # Q925P2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 26367
Test Specificity Detects mouse CEACAM-2 in direct ELISAs. Stains mouse CEACAM-2 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.