Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CEACAM-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB9214SP
This item is not returnable.
View return policy
Description
CEACAM2 Monoclonal specifically detects CEACAM2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CEACAM2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
26367 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Flow Cytometry, CyTOF | |
935610 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q925P2 | |
Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2 Gln35-Asp422 Accession # Q925P2 | |
25 μg | |
Primary | |
Detects mouse CEACAM-2 in direct ELISAs. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction