Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cerberus 1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$188.28 - $686.23
Specifications
| Antigen | Cerberus 1 |
|---|---|
| Clone | 225807 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
Cerberus 1 Monoclonal specifically detects Cerberus 1 in Mouse samples. It is validated for Western Blot.Specifications
| Cerberus 1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| O55233 | |
| 9350 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cerberus 1 Asp18-Pro272 Accession # O55233 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Cerberus 1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human Cerberus, recombinant mouse (rm) DAN, or rmGremlin. |
| 225807 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CER1, cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951 | |
| CER1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title