Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Ceruloplasmin ELISA Kit (Colorimetric)
SDP

Catalog No. NB398246 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398246 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398246 Supplier Novus Biologicals™ Supplier No. NBP282160
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 3.13 to 200 ng/mL
Assay Sensitivity 1.88 ng/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Supernatant
Specificity This kit recognizes Mouse Ceruloplasmin in samples. No Significant cross-reactivity or interference between Mouse Ceruloplasmin and analogues was observed.
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 1356
Gene Symbol CP
Quantity 1 Kit
Research Discipline Cardiovascular Biology, Neurodegeneration, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym ceruloplasmin, ceruloplasmin (ferroxidase), CP-2, EC 1.16.3.1, Ferroxidase
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.