Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Chemerin Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM2325
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM2325 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM2325 Supplier R&D Systems Supplier No. BAM2325
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Chemerin Monoclonal specifically detects Chemerin in Mouse samples. It is validated for ELISA Detection (Matched Antibody Pair).

Specifications

Antigen Chemerin
Applications ELISA
Classification Monoclonal
Clone 372402
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. Q9DD06
Gene Alias chemerin, RARRES2, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG-2, TIG2HP10433
Gene Symbols RARRES2
Host Species Rat
Immunogen E. coli-derived recombinant mouse Chemerin Thr17-Ser156 Accession # Q9DD06
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5919
Test Specificity Detects mouse Chemerin in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human Chemerin, recombinant mouse (rm) Cystatin C, or rmFetuin A is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.