Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Chemerin Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2325
This item is not returnable.
View return policy
Description
Chemerin Polyclonal specifically detects Chemerin in Mouse samples. It is validated for Western Blot.Specifications
Chemerin | |
Polyclonal | |
Biotin | |
Q9DD06 | |
RARRES2 | |
E. coli-derived recombinant mouse Chemerin Thr17-Ser156 Accession # Q9DD06 | |
50 μg | |
Primary | |
Detects mouse Chemerin in Western blots. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
chemerin, RARRES2, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG-2, TIG2HP10433 | |
Goat | |
Affinity Purified | |
RUO | |
5919 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction