Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Chitinase 3-like 1/YKL-40 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2649SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2649SP 25 μg
AF2649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2649SP Supplier R&D Systems Supplier No. AF2649SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Chitinase 3-like 1 Polyclonal specifically detects Chitinase 3-like 1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Chitinase 3-like 1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61362
Gene Alias AW208766, Brp39, Chi3l1, Chitinase 3 like 1, gp39, HCgp39, YKL-40
Gene Symbols Chil1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Chitinase 3-like 1 Tyr22-Ala381 Accession # Q61362
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1116
Test Specificity Detects mouse Chitinase 3-like 1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human Chitinase 3-like 1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.