Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Chitotriosidase/CHIT1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Chitotriosidase/CHIT1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Chitotriosidase/CHIT1 Polyclonal specifically detects Chitotriosidase/CHIT1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Chitotriosidase/CHIT1 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
CHI3, CHIT1, CHITCHI3, chitinase 1 (chitotriosidase), Chitinase-1, chitotriosidase-1, EC 3.2.1.14, FLJ00314, MGC125322, plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase | |
CHIT1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q9D7Q1 | |
1118 | |
Mouse myeloma cell line NS0-derived recombinant mouse Chitotriosidase/CHIT1 Ala22-Ser464 Accession # Q9D7Q1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Chitotriosidase/CHIT1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human CHIT1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title