Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CHL-1/L1CAM-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2147SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2147SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2147SP Supplier R&D Systems Supplier No. AF2147SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 12 publications

CHL-1/L1CAM-2 Polyclonal specifically detects CHL-1/L1CAM-2 in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen CHL-1/L1CAM-2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. BAC30699
Gene Alias CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein
Gene Symbols CHL1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CHL-1/L1CAM-2 Ala25-Gln1043 (Leu227-Gln242 del, Ala243Ser) Accession # BAC30699
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10752
Test Specificity Detects mouse CHL-1/L1CAM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human CHL-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.