Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Chordin Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB758SP
This item is not returnable.
View return policy
Description
Chordin Monoclonal specifically detects Chordin in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| Chordin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHRD, MGC133038, Sog | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8646 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA | |
| 119721 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9Z0E2 | |
| CHRD | |
| Mouse myeloma cell line NS0-derived recombinant mouse Chordin Thr27-Ser948 Accession # Q9Z0E2 | |
| 25 μg | |
| Primary | |
| Detects mouse Chordin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Pref-1 or rmWIF-1 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction