Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Chordin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF758
This item is not returnable.
View return policy
Description
Chordin Polyclonal specifically detects Chordin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
Chordin | |
Polyclonal | |
Unconjugated | |
Q9Z0E2 | |
CHRD | |
Mouse myeloma cell line NS0-derived recombinant mouse Chordin Thr27-Ser948 Accession # Q9Z0E2 | |
100 μg | |
Primary | |
Detects mouse Chordin in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
CHRD, MGC133038, Sog | |
Goat | |
Affinity Purified | |
RUO | |
8646 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction