Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CLEC4F/CLECSF13 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2784
This item is not returnable.
View return policy
Description
CLEC4F/CLECSF13 Monoclonal specifically detects CLEC4F/CLECSF13 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| CLEC4F/CLECSF13 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P70194 | |
| CLEC4F | |
| Mouse myeloma cell line NS0-derived recombinant mouse CLEC4F/CLECSF13 Ala65-Gly548 Accession # P70194 | |
| 100 μg | |
| Primary | |
| Detects mouse CLEC4F/CLECSF13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CLECSF4F, rhCLEC4D, rhCLEC4E, recombinant mouse (rm) OCIL, and rmOCIL-rp2 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 370901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLECSF13, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13, C-type lectin 13, C-type lectin domain family 4, member F, Galactose Particle Receptor, KCLR, KCR | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 165530 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction