Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CLEC4F/CLECSF13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2784SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2784SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2784SP Supplier R&D Systems Supplier No. MAB2784SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CLEC4F/CLECSF13 Monoclonal specifically detects CLEC4F/CLECSF13 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CLEC4F/CLECSF13
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 370901
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P70194
Gene Alias CLECSF13, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13, C-type lectin 13, C-type lectin domain family 4, member F, Galactose Particle Receptor, KCLR, KCR
Gene Symbols CLEC4F
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CLEC4F/CLECSF13 Ala65-Gly548 Accession # P70194
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165530
Test Specificity Detects mouse CLEC4F/CLECSF13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CLECSF4F, rhCLEC4D, rhCLEC4E, recombinant mouse (rm) OCIL, and rmOCIL-rp2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.