Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CLEC9a Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB67762G100UG
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal specifically detects CLEC9a in Mouse samples. It is validated for Flow Cytometry.Specifications
CLEC9a | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD370, C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CLEC9a, Lys57-Ile264, Accession # EDK99924 | |
100 μg | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
2456A | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
EDK99924 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
283420 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction