Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CLEC9a Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB67762SP Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB67762SP 25 μg
MAB67762100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB67762SP Supplier R&D Systems Supplier No. MAB67762SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CLEC9a Monoclonal specifically detects CLEC9a in Mouse samples. It is validated for CyTOF∣Flow Cytometry.

Specifications

Antigen CLEC9a
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 2456A
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. EDK99924
Gene Alias CD370, C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CLEC9a, Lys57-Ile264, Accession # EDK99924
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283420
Test Specificity Detects mouse CLEC9a in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.