Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Coagulation Factor III/Tissue Factor Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF3178
This item is not returnable.
View return policy
Description
Coagulation Factor III/Tissue Factor Polyclonal specifically detects Coagulation Factor III/Tissue Factor in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready.Specifications
Coagulation Factor III/Tissue Factor | |
Polyclonal | |
Unconjugated | |
P20352 | |
F3 | |
Mouse myeloma cell line NS0-derived recombinant mouse Coagulation Factor III/Tissue Factor Ala29-Glu251 Accession # P20352 | |
100 μg | |
Primary | |
Detects mouse Coagulation Factor III in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready | |
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
Goat | |
Affinity Purified | |
RUO | |
2152 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction