Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Coagulation Factor XIV/Protein C Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | Coagulation Factor XIV/Protein C |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Coagulation Factor XIV/Protein C Polyclonal specifically detects Coagulation Factor XIV/Protein C in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Coagulation Factor XIV/Protein C | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC, PROC1, Protein C, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C | |
PROC | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P33587 | |
5624 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Coagulation Factor XIV/Protein C Ile19-Leu460 Accession # P33587 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Coagulation Factor XIV/Protein C in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human Protein C is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title