Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse COCO Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3356
This item is not returnable.
View return policy
Description
COCO/DAND5 Monoclonal specifically detects COCO/DAND5 in Mouse samples. It is validated for Western Blot.Specifications
COCO/DAND5 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q76LW6 | |
DAND5 | |
E. coli-derived recombinant mouse COCO Arg24-Leu185 Accession # Q76LW6 | |
100 μg | |
Primary | |
Detects mouse COCO in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) COCO, rhDAN, recombinant mouse (rm) Gremlin, rmCerberus, and recombinant chicken Caronte is observed. | |
Mouse | |
Purified |
Western Blot | |
340531 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
199699 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction