Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Collagen XIII alpha 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4627SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4627SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4627SP Supplier R&D Systems Supplier No. AF4627SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Collagen XIII alpha 1 Polyclonal specifically detects Collagen XIII alpha 1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Collagen XIII alpha 1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH34164
Gene Alias COL13A1, collagen alpha-1(XIII) chain, collagen, type XIII, alpha 1, FLJ42485
Gene Symbols COL13A1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Collagen XIII alpha 1 Glu107-Gln565 Accession # AAH34164
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1305
Test Specificity Detects mouse Collagen XIII alpha 1 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human (rh) COL13A1v4 is observed, and less than 1% cross-reactivity with rhCOL4A1, rhCOL25A1, rhCOL3A1, and rhCOL1A1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.