Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Collectrin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4965
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4965 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4965 Supplier R&D Systems Supplier No. AF4965
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Collectrin/TMEM27 Polyclonal specifically detects Collectrin/TMEM27 in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Collectrin/TMEM27
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q9ESG4
Gene Alias collectrin, kidney-specific membrane protein, NX17, NX-17, TMEM27, transmembrane protein 27NX170610008J07Rik
Gene Symbols TMEM27
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Collectrin Glu15-Ile144 Accession # Q9ESG4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57393
Test Specificity Detects mouse Collectrin in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human Collectrin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.