Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Common gamma Chain/IL-2 R gamma Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7842
This item is not returnable.
View return policy
Description
Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Common gamma Chain/IL-2 R gamma | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P34902 | |
Il2rg | |
S. frugiperda insect ovarian cell line Sf21-derived recombinant mouse Common gamma Chain/IL-2 R gamma Trp23-Ala263 (predicted) Accession # P34902 | |
100 μg | |
Primary | |
Detects mouse Common gamma Chain/IL-2 R gamma in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-2 R alpha , rmIL-2 R beta , recombinant human (rh) IL-2 R gamma , rhIL-15 R alpha , recombinant rat IL-2 R alpha , or recombinant canine IL-2 R alpha is observed. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
644346 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 R gamma, IL-2 receptor subunit gamma, IL2R gamma, IL-2R subunit gamma, IL2RG, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3561 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction